Skip to Content
Merck
All Photos(1)

Key Documents

C2988

Sigma-Aldrich

Cytosol from Liver, Pooled

from human

Synonym(s):

Liver Cytosol

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$143.00

$143.00


Check Cart for Availability


Select a Size

Change View
1 VIAL
$143.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

$143.00


Check Cart for Availability

biological source

human

Quality Level

form

buffered aqueous solution

packaging

vial of ~20 mg (protein)

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

Application

Cytosolic preparations such as this may be used for screening potential substrates, inhibitors, and drugs to determine phase II drug metabolizing enzyme interactions.

Other Notes

Cytosol prepared from adult human donors. Contains measurable levels of NAT, N-acetyl transferase; GST, glutathione S-transferase; SULT, sulfotransferase.

Physical form

The product is supplied in a buffered solution containing 150 mM potassium chloride and 50 mM Tris, pH 7.5, with 2 mM EDTA.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service