Skip to Content
Merck
All Photos(1)

Key Documents

C2769

Sigma-Aldrich

Cobalt(II) nitrate hexahydrate

ACS reagent, 98.0-102.0%

Synonym(s):

Cobaltous nitrate, Cobaltous nitrate hexahydrate, Nitric acid, cobalt(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Co(NO3)2 · 6H2O
CAS Number:
Molecular Weight:
291.03
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

ACS reagent

Assay

98.0-102.0%

impurities

≤0.01% Insoluble matter

mp

55 °C (lit.)

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.005%
Cu: ≤0.002%
Fe: ≤0.001%
K: ≤0.01%
Mg: ≤0.005%
NH4+: ≤0.2%
Na: ≤0.05%
Ni: ≤0.15%
Pb: ≤0.002%
Zn: ≤0.01%

SMILES string

O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Co.2NO3.6H2O/c;2*2-1(3)4;;;;;;/h;;;6*1H2/q+2;2*-1;;;;;;

InChI key

QGUAJWGNOXCYJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William Buchmann et al.
Journal of mass spectrometry : JMS, 42(4), 517-526 (2007-02-27)
The electronebulization of a cobalt(II)/cysteine(Cys) mixture in water/methanol (50/50) produced mainly cobalt-cationized species. Three main groups of the Co-cationized species can be distinguished in the ESI-MS spectrum: (1) the cobalt complexes including the cysteine amino acid only (they can be
H H Chen et al.
Nucleic acids research, 10(11), 3561-3571 (1982-06-11)
Transient electric dichroism measurements have been used to observe the rotational relaxation times of 145 base pair fragments of poly (dGm5dC) and random sequence DNA to solution. From these the lengths of the fragments are calculated and the interbase pair
A time-resolved in situ quick-XAS investigation of thermal activation of Fischer-Tropsch silica-supported cobalt catalysts.
Jingping Hong et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(10), 2802-2805 (2012-02-07)
N G Al-Tawil et al.
Acta dermato-venereologica, 64(3), 203-208 (1984-01-01)
Peripheral blood lymphocytes from 45 patients with a positive patch test to cobalt chloride (CoCl2) and 37 controls were stimulated with various concentrations of CoCl2 and/or cobalt sulphate (CoSO4) or cobalt nitrate (Co(NO3)2) or cobalt acetate [CH3COO)2Co) for various days
J M Llobet et al.
Revista espanola de fisiologia, 39(3), 291-298 (1983-09-01)
The acute toxicities of chloride, acetate, cobalt nitrate and cobalt sulphate (II) were investigated in rats. The values obtained for the LD50 (7 days) were 133, 194, 198 and 279 mg of Co/kg respectively, when the salts were given orally.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service