Skip to Content
Merck
All Photos(1)

Key Documents

C-27115

Sigma-Aldrich

Chondrocyte Growth Medium

Basal Medium, phenol red-free, 500 ml

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$636.00

$636.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
$636.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

$636.00


Check Cart for Availability

Request a Bulk Order

form

solution

packaging

pkg of 500 ml

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Chondrocyte Basal Medium, phenol red-free (supplements sold separately)

Application

Chondrocyte Growth Medium has been developed for the in vitro cultivation of human chondrocytes. The fetal calf serum in the medium is strictly pretested for optimal performance. The medium is optimized for primary human cells, but can also be used for primary porcine and rat chondrocytes.Recommended for:Human Chondrocytes (HCH)

Components

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials

Quality

All lots of PromoCell Chondrocyte Growth Medium are subjected to comprehensive quality control tests using primary human chondrocytes. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested chondrocytes. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Storage and Stability

Store the Basal Medium at 4 to 8°C in the dark immediately after arrival. Do not freeze the Basal Medium. If stored properly, the medium is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C

Subculture Routine

Click here for more information.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Primary human chondrocyte cells and optimized growth media for the in vitro cultivation of chondrocytes. Protocols for handling, cell passaging, media, and product use.

Medium 199 supports non-transformed cell cultivation in virology and vaccine production, offering broad species applicability.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service