Skip to Content
Merck
All Photos(1)

Documents

C2646

Sigma-Aldrich

Copper(II) nitrate hydrate

Synonym(s):

Cupric nitrate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(NO3)2 · xH2O
CAS Number:
Molecular Weight:
187.56 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

mp

114.5 °C (dec.) (lit.)

SMILES string

O.[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Cu.2NO3.H2O/c;2*2-1(3)4;/h;;;1H2/q+2;2*-1;

InChI key

VMKYLARTXWTBPI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1 - Ox. Sol. 1 - Skin Corr. 1B

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shenghong Zhong et al.
Soft matter, 11(35), 6999-7004 (2015-08-05)
Here, we report the development of an experimental system based on liquid crystals (LCs) confined in microcapillaries for imaging interfacial phenomena. The inner surfaces of the microcapillaries were modified with octadecyltrichlorosilane to promote an escaped-radial configuration of LCs. We checked

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service