Skip to Content
Merck
All Photos(1)

Key Documents

C2499

Sigma-Aldrich

CP-339818

≥98% (HPLC)

Synonym(s):

N-[1-(phenylmethyl)-4(1H)-quinolinylidene]-1-pentanamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$119.00
25 MG
$477.00

$119.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$119.00
25 MG
$477.00

About This Item

Empirical Formula (Hill Notation):
C21H24N2 · HCl
CAS Number:
Molecular Weight:
340.89
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$119.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >10 mg/mL

originator

Wyeth

storage temp.

2-8°C

SMILES string

Cl.CCCCC\N=C1/C=CN(Cc2ccccc2)c3ccccc13

InChI

1S/C21H24N2.ClH/c1-2-3-9-15-22-20-14-16-23(17-18-10-5-4-6-11-18)21-13-8-7-12-19(20)21;/h4-8,10-14,16H,2-3,9,15,17H2,1H3;1H/b22-20+;

InChI key

JIRISCAPZWLWCG-QPNALZDCSA-N

Biochem/physiol Actions

CP-338818 blocks both Kv1.3 and Kv1.4. Kv1.3 is expressed in brain and in effector memory T (Tem) cells, and Kv1.4 is expressed in brain cells.
CP-338818 blocks both Kv1.3 and Kv1.4. Kv1.3 is expressed in brain and in effector memory T (Tem) cells, and Kv1.4 is expressed in brain cells. Kv1.3 and Kv1.4 are members of the Shaker family of voltage-gated potassium channels. Both channels are involved in setting the membrane potential of neurons. In addition, Kv1.3 maintains the membrane potential for T memory cells and is up-regulated upon T cell activation, contributing to multiple immune processes and disorders. Blockers of Kv1.3 have long been sought for therapeutic benefit, and they are also valuable tools for studying the immune system. CP-339818 is valuable for immune system studies, where the absence of Kv1.4 makes it functionally selective for Kv1.3, and it has shown suppression of T cell activation. CP-339818 is also valuable for the study of Kv1.4 function.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Wyeth. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandre J C Loucif et al.
British journal of pharmacology, 175(12), 2272-2283 (2017-11-19)
TREK two-pore-domain potassium (K2P ) channels play a critical role in regulating the excitability of somatosensory nociceptive neurons and are important mediators of pain perception. An understanding of the roles of TREK channels in pain perception and, indeed, in other

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service