Pericytes are positive for CD146, PDGFRβ, αSMA, CD13, NG2, Desmin, and negative for CD31, CD34, CD45, and CD56.
C-14095
Human Pericytes from Placenta (hPC-PL)
1 million cells per pellet
Select a Size
About This Item
Recommended Products
biological source
human placenta
packaging
pkg of 1,000,000 cells
morphology
(pericyte)
application(s)
cell analysis (DNA, RNA, and protein)
shipped in
dry ice
storage temp.
−20°C
General description
Cell Line Origin
Application
Quality
Warning
Physical form
Subculture Routine
Other Notes
Legal Information
Disclaimer
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
What are pericytes? What are common pericyte markers? Are pericytes mesenchymal stem cells? Learn more about pericytes in our FAQ page. Explore over 350 PromoCell products.
Primary human pericytes and cell culture media optimized for the in vitro cultivation of human pericytes from all tissues. Protocols for handling, cell passaging, media, and product use.
Primary cells from PromoCell are available in pelleted form for research applications like gene expression analysis, and cell/protein characterization experiments.
-
What markers are used to characterize pericytes?
1 answer-
Helpful?
-
-
How long do pericytes grow in culture?
1 answer-
PromoCell’s human pericytes derived from microvessels of the human placenta are cryopreserved at the end of secondary culture (P1) and after thawing are considered P2. The number of populations doublings is not determined for each individual cell lot, however they are guarenteed to be grown for at least 15 population doublings.
Helpful?
-
-
How are pericytes isolated?
1 answer-
Pericyte isolation generally starts with enzymatic digestion of tissue with Trypsin EDTA, usually followed by the isolation of microvessel fragments via successive filtration steps. Pure pericyte cultures can be obtained through either positive or negative immuno-selection via the use of magnetic beads or flow cytometry using the antibody markers above.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service