Skip to Content
Merck
All Photos(1)

Key Documents

C-14080

Sigma-Aldrich

Human Cardiac Myocytes (HCM)

1 million cells per pellet

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$770.00

$770.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$770.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

$770.00


Check Cart for Availability

Request a Bulk Order

biological source

human heart (ventricles)

packaging

pkg of 1,000,000 cells

morphology

(myocytes)

application(s)

cell analysis (DNA, RNA, and protein)

shipped in

dry ice

storage temp.

−20°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details. Cells are dissolved in RNAlater for subsequent RNA, DNA, or protein analysis. Note that cell pellets cannot be revived.

Cell Line Origin

Heart

Application

Primary Human Cardiac Myocytes (HCM) are isolated from the ventricles of the adult heart. They are qualified for in vitro research on cardiac diseases and for pharmacological studies. Unlike freshly isolated rod-shaped myocytes, cultured HCM can be used for long-term experiments like investigating the long-term effects of cytokines, mechanical strain, or cell-cell interactions, as they are prepared according to a special protocol.Initially, the HCM act more like progenitor cells in that they are not yet fully differentiated. They express the markers of early stage differentiation such as GATA-4 and sarcomeric alpha-actin and have a high capacity for proliferation. When they are grown to confluency and cultivated for an extended period of time, the differentiation process begins. Markers of late differentiation (e.g. sarcomeric alpha-actinin, slow muscle myosin) are increased and the cells begin to form myotube-like structures.

Quality

Rigid quality control tests are performed for each lot of the corresponding cells. They are tested for cell viability, cell morphology, growth performance and for cell-type specific markers. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Warning

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.

Physical form

Pellet is dissolved in RNAlater® for downstream analysis of DNA, RNA or protein profiles. Cells are not revivable.

Subculture Routine

Click here for more information.

Other Notes

The Cell Pellets can be stored indefinitely at -20°C, and for up to 1 month at 4°C. Note: The Cell Pellets will not freeze at -20°C. Always centrifuge product briefly before opening vial.

Legal Information

RNAlater is a registered trademark of Ambion, Inc.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Primary human cardiac myocytes isolated from normal human ventricle tissue of the adult heart, plus optimized myocyte media. Protocols for handling, cell passaging, media, and product use.

Primary cells from PromoCell are available in pelleted form for research applications like gene expression analysis, and cell/protein characterization experiments.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service