C1244
CHM-1 hydrate
≥99% (HPLC), solid
Synonym(s):
2-(2-fluorophenyl)-6,7-methylenedioxy-2-4-quinolone hydrate, NSC 656158
About This Item
Recommended Products
Assay
≥99% (HPLC)
form
solid
storage condition
desiccated
solubility
DMSO: ≥5 mg/mL
storage temp.
2-8°C
SMILES string
O.Fc1ccccc1C2=CC(=O)c3cc4OCOc4cc3N2
InChI
1S/C16H10FNO3.H2O/c17-11-4-2-1-3-9(11)12-6-14(19)10-5-15-16(21-8-20-15)7-13(10)18-12;/h1-7H,8H2,(H,18,19);1H2
InChI key
KIAVGSCBJYDTIM-UHFFFAOYSA-N
Biochem/physiol Actions
Features and Benefits
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Review properties, activators and inhibitors, and available products for researching cyclin-dependent kinases (CDKs).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service