Skip to Content
Merck
All Photos(1)

Documents

C0606

Sigma-Aldrich

Protein Extraction Reagent Type 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.56

General description

Protein Extraction Reagent Type 2 is a highly effective protein solubilization and extraction reagent composed urea, thiourea, Trizma®, CHAPS, and detergent SB3-10.

Application

Premixed and prequalified, Protein Extraction Reagent Type 2 is a highly effective protein solubilization and extraction reagent ideal for protein extraction prior to isoelectric focusing (IEF) and 2D electrophoresis. Reducing and alkylating reagents TBP (cat# T7567) and iodoacetamide (cat# A3221) are recommended for use with this reagent. Protein Extraction Reagent Type 2 is also available as a component of the ProteoPrep® Sample Extraction Kit (PROT-TOT).

Components

Contains urea, thiourea, Trizma®, CHAPS, and detergent SB3-10.

Legal Information

ProteoPrep is a registered trademark of Merck KGaA, Darmstadt, Germany
Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sumihisa Orita et al.
BMC musculoskeletal disorders, 12, 134-134 (2011-06-18)
The exact mechanism of knee osteoarthritis (OA)-associated pain is unclear, whereas mixed evidence of inflammatory pain and neuropathic pain has been noted. We aimed to investigate pain-related sensory innervation in a monoiodoacetate (MIA)-induced model of OA. Sixty of seventy female
Xiaoping Ao et al.
Journal of hematology & oncology, 2, 6-6 (2009-02-04)
Recent research has supported that a variety of cytokines play important roles during radiation-induced lung toxicity. The present study is designed to investigate the differences in early cytokine induction after radiation in sensitive (C57BL/6) and resistant mice (C3H). Twenty-two cytokines

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service