Key Documents
BMSIPS2050PS3
Sigma-Aldrich
Benchmark Overhead Stirrer Propeller
flat blade, stainless steel
Select a Size
1 EACH
$161.00
$161.00
Check Cart for Availability
Select a Size
Change View
1 EACH
$161.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.72
$161.00
Check Cart for Availability
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific IPS2050-P-S1
diam. × L
68 mm × 400 mm
General description
Stiring rods and propellers to fit Benchmark range of OS-20L and OS-40L overhead stirrers.
Useful for gentle, mid to low speed mixing, this stainless steel flat propeller has 3 holes.
Antibody Adsorbant
stirring, mixing
Choose from one of the most recent versions:
Certificates of Analysis (COA)