Skip to Content
Merck
All Photos(1)

Key Documents

BM0015

Sigma-Aldrich

BMS-309403

≥98% (HPLC)

Synonym(s):

2-[[2′-(5-Ethyl-3,4-diphenyl-1H-pyrazol-1-yl)[1,1′-biphenyl]-3-yl]oxy]acetic acid, BMS 309403

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H26N2O3
CAS Number:
Molecular Weight:
474.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

SMILES string

O=C(O)COC1=CC(C2=CC=CC=C2N3C(CC)=C(C4=CC=CC=C4)C(C5=CC=CC=C5)=N3)=CC=C1

Biochem/physiol Actions

BMS-309403 is a potent and selective inhibitor of fatty acid binding protein 4 (FABP4), also known as adipocyte FABP (A-FABP, aP2). FABP4 is an intracellular lipid-binding protein responsible for the transportation of fatty acids. It is expressed primarily in adipose tissue and is associated with inflammation, obesity, diabetes and cardiovascular diseases. BMS309403 interacts with the fatty-acid-binding pocket within the interior of FABP4 and has been shown to competitively inhibit fatty acid binding with a Ki value < 2 nM. BMS309403 is orally active and has been shown to reduce atherosclerosis in mice studies.
BMS-309403 is a potent and selective inhibitor of fatty acid binding protein 4 (FABP4).

Features and Benefits

BMS-309403 is available through a partnership with Bristol-Myers Squibb (BMS). To learn more and view other BMS compounds, visit sigma.com/BMS.

Legal Information

Sold for research purposes only under agreement from BMS.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammed Haider et al.
Biomedicines, 9(11) (2021-11-28)
Atherosclerosis is a chronic degenerative disorder characterized by lipid-dense plaques and low-grade inflammation affecting arterial walls. Foamy macrophages are important in the formation of atherosclerotic plaques and the induction of low-grade inflammation. The presence of lipid-laden macrophages has occurred in
Yuxian Guo et al.
Nature communications, 12(1), 7094-7094 (2021-12-09)
Oxidative stress contributes to the pathogenesis of acute lung injury. Protein S-glutathionylation plays an important role in cellular antioxidant defense. Here we report that the expression of deglutathionylation enzyme Grx1 is decreased in the lungs of acute lung injury mice.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service