Skip to Content
Merck
All Photos(1)

Documents

B9811

Sigma-Aldrich

PolyTransport Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

liquid

pH

7.2-7.6

application(s)

hematology
histology

storage temp.

room temp

General description

PolyTransport Buffer is a medium used to transport and preserve specimens from surgery to the point of grossing for fixation prior to further processing. The specially formulated transport medium is designed to preserve tissue in a viable state during transport by maintaining osmolarity and pH for 24 to 48 hours.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service