B8148
6-Benzylaminopurine 9-(β-D-glucoside)
Synonym(s):
N6-Benzyladenine 9-glucoside
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C18H21N5O5
CAS Number:
Molecular Weight:
387.39
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
Recommended Products
storage temp.
−20°C
SMILES string
OCC1OC(C(O)C(O)C1O)n2cnc3c(NCc4ccccc4)ncnc23
General description
Major metabolite of the cytokinin, 6-benzylaminopurine (BAP).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service