Skip to Content
Merck
All Photos(1)

Key Documents

B7563

Sigma-Aldrich

BRD7389

≥98% (HPLC)

Synonym(s):

1-Phenethylamino-3H-naphtho[1,2,3-de]quinoline-2,7-dione, 1-[(2-Phenylethyl)amino]-naphtho-3H-[1,2,3-de]quinoline-2,7-dione

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$273.00

$273.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$273.00

About This Item

Empirical Formula (Hill Notation):
C24H18N2O2
CAS Number:
Molecular Weight:
366.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$273.00


Estimated to ship on16 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

orange

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

[nH]1c2c3c(c5c([c](c3ccc2)=O)cccc5)c([c]1=O)NCCc4ccccc4

InChI

1S/C24H18N2O2/c27-23-17-10-5-4-9-16(17)21-20-18(23)11-6-12-19(20)26-24(28)22(21)25-14-13-15-7-2-1-3-8-15/h1-12,25H,13-14H2,(H,26,28)

InChI key

XASCINRGTHLHGM-UHFFFAOYSA-N

Biochem/physiol Actions

BRD7389 is an inhibitor of the RSK kinase family.
BRD7398 is a RSK kinase inhibitor that induces insulin expression and differentiation of pancreatic alpha into beta cells. BRD7398 inhibits RSK1, RSK2 and RSK3; IC50 values of 1.5, 2.4 and 1.2 uM, respectively. Treatment of mouse aTC1 cells with BRD7398 induced expression of b-cell markers Pdx1, Pax4, Iapp and Npy and also induced insulin production.

Features and Benefits

This compound is featured on the MAPKAPs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dana M Gwinn et al.
Cancer cell, 33(1), 91-107 (2018-01-10)
KRAS is a regulator of the nutrient stress response in non-small-cell lung cancer (NSCLC). Induction of the ATF4 pathway during nutrient depletion requires AKT and NRF2 downstream of KRAS. The tumor suppressor KEAP1 strongly influences the outcome of activation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service