Skip to Content
Merck
All Photos(1)

Key Documents

B7188

Sigma-Aldrich

Blonanserin

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$84.00
25 MG
$320.00

$84.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$84.00
25 MG
$320.00

About This Item

Empirical Formula (Hill Notation):
C23H30FN3
CAS Number:
Molecular Weight:
367.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$84.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥10 mg/mL

originator

Dainippon Sumitomo

storage temp.

room temp

SMILES string

CCN1CCN(CC1)c2cc(-c3ccc(F)cc3)c4CCCCCCc4n2

InChI

1S/C23H30FN3/c1-2-26-13-15-27(16-14-26)23-17-21(18-9-11-19(24)12-10-18)20-7-5-3-4-6-8-22(20)25-23/h9-12,17H,2-8,13-16H2,1H3

InChI key

XVGOZDAJGBALKS-UHFFFAOYSA-N

Biochem/physiol Actions

Blonanserin is a D2/5-HT2 receptor antagonist; atypical antipsychotic.

Features and Benefits

This compound was developed by Dainippon Sumitomo. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quetiapine-induced insulin resistance after switching from blonanserin despite a loss in both bodyweight and waist circumference.
Yutaro Suzuki et al.
Psychiatry and clinical neurosciences, 66(6), 534-535 (2012-10-17)
Ming Peng et al.
Journal of chromatographic science, 50(8), 727-732 (2012-05-19)
Blonanserin is a novel atypical antipsychotic agent for the treatment of schizophrenia. Ethyl alcohol, isopropyl alcohol and toluene are utilized in the synthesis route of this bulk drug. A new validated gas chromatographic (GC) method for the simultaneous determination of
Koji Kato et al.
Psychiatry and clinical neurosciences, 65(4), 389-391 (2011-05-17)
The purpose of the present study was to provide preliminary data on the usefulness and safety of blonanserin for patients with delirium in the intensive care unit (ICU). The charts of 32 consecutive patients with delirium in the ICU were
Three cases of schizophrenia showing improvement after switching to blonanserin.
Naohiro Kurotaki et al.
Psychiatry and clinical neurosciences, 65(4), 396-397 (2011-06-21)
Tomomi Tenjin et al.
Human psychopharmacology, 27(1), 90-100 (2012-01-27)
The purpose of this study was to evaluate the effects of blonanserin, a novel antipsychotic, on cognitive function in first-episode schizophrenia. Twenty-four antipsychotic-naïve patients with first-episode schizophrenia participated in the study. Blonanserin was given in an open-label design for 8 weeks.

Articles

We offer many products related to serotonin receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service