Skip to Content
Merck
All Photos(1)

Key Documents

B6893

Sigma-Aldrich

Benzyl 2-acetamido-2-deoxy-β-D-glucopyranoside

Synonym(s):

β-D-GlcNAc-1→OCH2Ph, Benzyl N-acetyl-β-D-glucosaminide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO6
CAS Number:
Molecular Weight:
311.33
MDL number:
UNSPSC Code:
12352201

storage temp.

−20°C

SMILES string

CC(=O)NC1C(O)C(O)C(CO)OC1OCc2ccccc2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang Di et al.
Insect biochemistry and molecular biology, 84, 48-62 (2017-04-04)
Lepidopteran caterpillars rely on olfaction and gustation to discriminate among food sources. Compared to the larval gustation, the larval olfaction has been poorly investigated. To uncover the molecular basis of olfaction in Helicoverpa armigera larvae, we identified 17 odorant receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service