Skip to Content
Merck
All Photos(1)

Documents

B6769

Sigma-Aldrich

des-Arg9-[Leu8]-Bradykinin acetate salt

≥97% (HPLC)

Synonym(s):

Arg-Pro-Pro-Gly-Phe-Ser-Pro-Leu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H63N11O10
CAS Number:
Molecular Weight:
870.01
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

des-Arg9-[Leu8]-Bradykinin acetate salt, ≥97% (HPLC)

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

CC(O)=O.CC(C)C[C@H](NC(=O)[C@@H]1CCCN1C(=O)[C@H](CO)NC(=O)[C@H](Cc2ccccc2)NC(=O)CNC(=O)[C@@H]3CCCN3C(=O)[C@@H]4CCCN4C(=O)[C@@H](N)CCCNC(N)=N)C(O)=O

InChI

1S/C41H63N11O10.C2H4O2/c1-24(2)20-28(40(61)62)48-36(57)31-14-8-17-50(31)38(59)29(23-53)49-34(55)27(21-25-10-4-3-5-11-25)47-33(54)22-46-35(56)30-13-7-18-51(30)39(60)32-15-9-19-52(32)37(58)26(42)12-6-16-45-41(43)44;1-2(3)4/h3-5,10-11,24,26-32,53H,6-9,12-23,42H2,1-2H3,(H,46,56)(H,47,54)(H,48,57)(H,49,55)(H,61,62)(H4,43,44,45);1H3,(H,3,4)/t26-,27-,28-,29-,30-,31-,32-;/m0./s1

InChI key

XBFMNOKHQZCARQ-XXZMQFNDSA-N

Amino Acid Sequence

Arg-Pro-Pro-Gly-Phe-Ser-Pro-Leu

Application

The Bradkykinin amid peptide des-Arg9-[Leu8]-Bradykinin is used as a bradykinin-1 receptor (B1R) antagonist frequently in comparison with the B1 receptor agonist des-Arg9-bradykinin.

Biochem/physiol Actions

Bradykinin receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juliano Ferreira et al.
Neuropharmacology, 54(3), 597-604 (2008-01-01)
The peripheral injection of phorbol myristate acetate (PMA) into the mouse paw induces nociception mediated through activation of protein kinase C (PKC). In the present study, we examine the contribution of kinin B1 receptor to PMA-induced nociception. Nociception was assessed
Thomas A Barker et al.
Hypertension (Dallas, Tex. : 1979), 48(5), 942-949 (2006-09-20)
It has been suggested that the effects of angiotensin II type 1 receptor (AT1R) blockers are in part because of angiotensin II type 2 receptor (AT2R) signaling. Interactions between the AT2R and kinins modulate cardiovascular function. Because AT2R expression increases
M M Campos et al.
British journal of pharmacology, 117(5), 793-798 (1996-03-01)
1. The effect of pretreatment with bacterial endotoxin (LPS, 10 micrograms, i.v., 24 h) on the bradykinin B1 and B2 receptor-induced oedema in the rat paw, and the interaction of B1-mediated responses with other inflammatory mediators, was investigated. 2. Intraplantar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service