Skip to Content
Merck
All Photos(1)

Key Documents

B6385

Sigma-Aldrich

Benzoylated Naphthoylated DEAE–Cellulose

medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
47101511
NACRES:
NA.56

form

powder

Quality Level

particle size

, medium

storage temp.

2-8°C

Application

Benzoylated Naphthoylated DEAE-cellulose is a cellulose medium used in protein chromatography, ion exchange chromatography and anion exchange media. Benzoylated Naphthoylated DEAE is particularly useful for the separation of single and double stranded DNA. Benzoylated Naphthoylated DEAE has been used to design a more efficient approach for the detection of human papillomavirus DNA and to study the cytotoxcity of the drug 5-Fluorouracil (FUra).

Preparation Note

Prepared by the method of Gillam et al., Biochemistry, 6, 3043 (1967).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A N Koterov et al.
Biokhimiia (Moscow, Russia), 57(2), 195-200 (1992-02-01)
Single-stranded DNA-binding proteins (SSB-proteins) isolated from Ehrlich ascites tumour (EAT) cells were incubated for 30 min at 5 mM NaCl with salmon sperm DNA or [3H]DNA from EAT at the SSB-protein/DNA ratio (w/w) of 0 to 4.5. After addition of
J D Schuetz et al.
Cancer chemotherapy and pharmacology, 21(3), 208-210 (1988-01-01)
5-Fluorouracil (FUra) was previously demonstrated to be incorporated into DNA at cytotoxic concentrations in mouse bone marrow cells. Subsequently, we showed that under these conditions FUra exhibited a time-dependent removal from DNA accompanied by a decrease in DNA strand length.
R M Knobler et al.
Clinical and experimental dermatology, 17(6), 392-396 (1992-11-01)
Using the benzoylated naphthoylated DEAE cellulose method (BND-method) we have designed a more efficient approach for the detection of human papillomavirus-DNA (HPV-DNA) via dot-blot and hybridization. Biopsy material from anogenital warts (40 patients), invasive carcinoma uteri (12 patients) and normal
Michaela A Gold et al.
PLoS genetics, 17(10), e1009863-e1009863 (2021-10-22)
Disease-associated trinucleotide repeats form secondary DNA structures that interfere with replication and repair. Replication has been implicated as a mechanism that can cause repeat expansions and contractions. However, because structure-forming repeats are also replication barriers, it has been unclear whether
Ana Teixeira-Silva et al.
Nature communications, 8(1), 1982-1982 (2017-12-08)
Replication requires homologous recombination (HR) to stabilize and restart terminally arrested forks. HR-mediated fork processing requires single stranded DNA (ssDNA) gaps and not necessarily double strand breaks. We used genetic and molecular assays to investigate fork-resection and restart at dysfunctional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service