Skip to Content
Merck
All Photos(1)

Key Documents

B6311

Sigma-Aldrich

BTO-1

≥98% (HPLC), solid

Synonym(s):

5-Cyano-7-nitro-2-benzothiazolecarboxamide-3-oxide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$149.00
25 MG
$591.00

$149.00


Estimated to ship on24 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$149.00
25 MG
$591.00

About This Item

Empirical Formula (Hill Notation):
C9H4N4O4S
CAS Number:
Molecular Weight:
264.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$149.00


Estimated to ship on24 May 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

form

solid

color

tan

solubility

H2O: <2 mL/mL
DMSO: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

NC(=O)c1sc2c(cc(cc2[n+]1[O-])C#N)[N+]([O-])=O

InChI

1S/C9H4N4O4S/c10-3-4-1-5-7(6(2-4)13(16)17)18-9(8(11)14)12(5)15/h1-2H,(H2,11,14)

InChI key

FKRBAXZAJBBIAJ-UHFFFAOYSA-N

Biochem/physiol Actions

BTO-1 is a polo-like kinase (Plk) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Campbell McInnes et al.
Nature chemical biology, 2(11), 608-617 (2006-10-10)
Polo-like kinases (Plks) have several functions in mitotic progression and are upregulated in many tumor types. Small-molecule Plk inhibitors would be valuable as tools for studying Plk biology and for developing antitumor agents. Guided by homology modeling of the Plk1
Kristin P Leister et al.
Current chemical genomics, 5, 21-29 (2011-06-07)
Tumor Necrosis Factor-α (TNF-α), a secreted cytokine, plays an important role in inflammatory diseases and immune disorders, and is a potential target for drug development. The traditional assays for detecting TNF-α, enzyme linked immunosorbent assay (ELISA) and radioimmunoassay, are not
Mark E Burkard et al.
PLoS biology, 7(5), e1000111-e1000111 (2009-05-27)
Animal cells initiate cytokinesis in parallel with anaphase onset, when an actomyosin ring assembles and constricts through localized activation of the small GTPase RhoA, giving rise to a cleavage furrow. Furrow formation relies on positional cues provided by anaphase spindle
Keiko Yata et al.
Molecular cell, 45(3), 371-383 (2012-02-14)
Homologous recombination (HR) plays an important role in the maintenance of genome integrity. HR repairs broken DNA during S and G2 phases of the cell cycle but its regulatory mechanisms remain elusive. Here, we report that Polo-like kinase 1 (Plk1)
Conrad von Schubert et al.
PLoS biology, 8(9) (2010-10-12)
The protozoan parasite Theileria inhabits the host cell cytoplasm and possesses the unique capacity to transform the cells it infects, inducing continuous proliferation and protection against apoptosis. The transforming schizont is a multinucleated syncytium that resides free in the host

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service