Skip to Content
Merck
All Photos(1)

Documents

B5651

Sigma-Aldrich

Benzyldimethyltetradecylammonium chloride

Synonym(s):

Benzyltetradecyldimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13N(Cl)(CH3)2CH2C6H5
CAS Number:
Molecular Weight:
368.04
Beilstein:
4062599
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

description

cationic

mp

56-62 °C

storage temp.

2-8°C

SMILES string

[Cl-].CCCCCCCCCCCCCC[N+](C)(C)Cc1ccccc1

InChI

1S/C23H42N.ClH/c1-4-5-6-7-8-9-10-11-12-13-14-18-21-24(2,3)22-23-19-16-15-17-20-23;/h15-17,19-20H,4-14,18,21-22H2,1-3H3;1H/q+1;/p-1

InChI key

OCBHHZMJRVXXQK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service