Skip to Content
Merck
All Photos(1)

Key Documents

B5186

Sigma-Aldrich

Monoclonal Anti-BAF155 antibody produced in mouse

~2 mg/mL, clone DXD7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-BRG1-associated Factor 155 KL, Anti-SMARCC1, Anti-SWI/SNF-related, matrix-associated, actin-dependent regulator of chromatin, subfamily C, member 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

DXD7, monoclonal

form

buffered aqueous solution

mol wt

antigen ~155 kDa

species reactivity

human

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: 2-4 μg/mL using total cell extract of MCF7 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SMARCC1(6599)

Specificity

The antibody does not react with BAF170.

Immunogen

synthetic peptide corresponding to amino acids 591-608 of human BAF155, conjugated to KLH.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arsalan Shabbir et al.
American journal of physiology. Heart and circulatory physiology, 299(5), H1428-H1438 (2010-09-21)
We recently demonstrated a cardiac therapeutic regimen based on injection of bone marrow mesenchymal stem cells (MSCs) into the skeletal muscle. Although the injected MSCs were trapped in the local musculature, the extracardiac cell delivery approach repaired the failing hamster
Stephanie A Morris et al.
Nature structural & molecular biology, 21(1), 73-81 (2013-12-10)
ATP-dependent chromatin remodeling is an essential process required for the dynamic organization of chromatin structure. Here we describe the genome-wide location and activity of three remodeler proteins with diverse physiological functions in the mouse genome: Brg1, Chd4 and Snf2h. The
Eric Allemand et al.
PLoS genetics, 12(9), e1006318-e1006318 (2016-09-24)
Several studies propose an influence of chromatin on pre-mRNA splicing, but it is still unclear how widespread and how direct this phenomenon is. We find here that when assembled in vivo, the U2 snRNP co-purifies with a subset of chromatin-proteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service