Skip to Content
Merck
All Photos(1)

Documents

B4555

Sigma-Aldrich

Benazoline oxalate salt

solid

Synonym(s):

4,5-Dihydro-2-(2-naphthalenyl)-1H-imidazole oxalate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12N2 · C2H2O4
CAS Number:
Molecular Weight:
286.28
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

solubility

H2O: ≤8 mg/mL

SMILES string

OC(=O)C(O)=O.c1ccc2cc(ccc2c1)-c3ncc[nH]3

InChI

1S/C13H10N2.C2H2O4/c1-2-4-11-9-12(6-5-10(11)3-1)13-14-7-8-15-13;3-1(4)2(5)6/h1-9H,(H,14,15);(H,3,4)(H,5,6)

InChI key

RIKYOUPROWKIIF-UHFFFAOYSA-N

Biochem/physiol Actions

I2 imidazoline receptor agonist; has high affinity and selectivity for I2 imidazoline receptors over α-adrenoceptors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Pigini et al.
Bioorganic & medicinal chemistry, 5(5), 833-841 (1997-05-01)
The observation that all the attempts to characterize imidazoline (I) receptors have been carried out with non-selective or poorly selective ligands prompted us to undertaken research aimed at developing selective ligand(s). In previous work using, as a starting point, cirazoline
H Greney et al.
Molecular pharmacology, 57(6), 1142-1151 (2000-05-29)
Clonidine and benazoline are two structurally related imidazolines. Whereas clonidine binds both to alpha(2)-adrenoceptors (alpha(2)R) and to I(1) imidazoline receptors (I(1)R), benazoline showed a high selectivity for imidazoline receptors. Although the alpha(2)R are negatively coupled to adenylate cyclase, no effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service