Skip to Content
Merck
All Photos(1)

Key Documents

B3680

Sigma-Aldrich

Monoclonal Anti-Bone Morphogenetic Protein 6 antibody produced in mouse

clone 74219.11, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-BMP-6

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$1,310.00

$1,310.00


Estimated to ship on20 May 2025



Select a Size

Change View
500 μG
$1,310.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

$1,310.00


Estimated to ship on20 May 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

74219.11, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

capture ELISA: suitable
neutralization: suitable
western blot: 1-2 μg/mL

isotype

IgG2b

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BMP6(654)

Related Categories

General description

Bone morphogenetic proteins (BMPs) are the members of the TGF-β superfamily and are identified as the critical factors for formation of bone and cartilage. Apart from this BMPs have immune regulatory functions. The cellular effects of BMPs are mediated by a heteromeric complex composed of type I and type II receptors. BMP-6 regulates both T cell and B cell-mediated immunity. It acts through Smad and p38 pathway to induce the expression of IL-6. Additionally, it induces the expression of iNOS by macrophages through the activation of NF-κB and STAT pathways. BMP-6 is a stimulator of differentiation of liver, bone, neuronal tissue and embryonic kidney and urinary systems. Studies also indicate that BMP-6 is important in the regulation of liver hormone, hepcidin, expression that is important to maintain systemic iron balance
Monoclonal Anti-Bone Morphogenetic Protein 6 recognizes human BMP-6. The antibody shows no cross-reactivity with recombinant human (rh) BMP-2, rhBMP-4, rhBMP-5, and rhBMP-7.

Immunogen

recombinant human BMP-6.

Application

Anti-Bone Morphogenetic Protein 6 antibody may be used for immunoblotting at a working concentration of 1.0-2.0 μg/ml. For capture ELISA a concentration of 4 μg/ml is recommemnded. The antibody is also suitable for neutralization reactions.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Preparation Note

Purified using protein G.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Hyuk Hong et al.
Immunology, 128(1 Suppl), e442-e450 (2009-02-05)
Bone morphogenetic proteins (BMPs) are members of the transforming growth factor (TGF)-beta superfamily which regulates bone formation, haematopoiesis and development. While TGF-beta is known to be a negative regulator of the immune system, the effect of BMPs on the immune
Seok J Kwon et al.
Immunology, 128(1 Suppl), e758-e765 (2009-09-25)
Bone morphogenetic proteins (BMPs) are members of the transforming growth factor-beta (TGF-beta) superfamily. In the present study, we investigated the effect of BMPs on the production of inducible nitric oxide synthase (iNOS) in the murine macrophage cell line, RAW 264.7
Elena Corradini et al.
Hepatology (Baltimore, Md.), 54(1), 273-284 (2011-04-14)
The bone morphogenetic protein 6 (BMP6)-SMAD signaling pathway is a central regulator of hepcidin expression and systemic iron balance. However, the molecular mechanisms by which iron is sensed to regulate BMP6-SMAD signaling and hepcidin expression are unknown. Here we examined
Geun Taek Lee et al.
The Journal of biological chemistry, 285(50), 39401-39408 (2010-10-05)
Unlike the prototype transforming growth factor-β (TGF-β), bone morphogenetic protein-6 (BMP-6) activates macrophages. Here, we report that BMP-6 induces the expression of IL-6 in macrophages. Using overexpression and knockdown experiments, we demonstrate that BMP receptor type II and activin-like kinase-2
Kohei Miyazono et al.
Cytokine & growth factor reviews, 16(3), 251-263 (2005-05-06)
Bone morphogenetic proteins (BMPs), members of the transforming growth factor-beta (TGF-beta) superfamily, bind to two different serine/threonine kinase receptors, and mediate their signals through Smad-dependent and Smad-independent pathways. Receptor regulated-Smad (R-Smad) proteins specific for the BMP pathways interact with various

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service