Skip to Content
Merck
All Photos(1)

Documents

B1682

Sigma-Aldrich

BAD from mouse

≥70% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in E. coli

Quality Level

description

full-length murine Bad with an N-terminal histidine-tag.

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

31 kDa

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Bad(12015)

Biochem/physiol Actions

Bad, a pro-apoptotic factor, is a member of the Bcl-2 family. Cytokine-induced phosphorylation of Bad results in it being sequested in the cytosol. In the absence of cytokines such as interleukin 3 (IL-3), Bad dephosphorylation occurs, causing the release of cytochrome c from the mitochondria.

Physical form

Supplied as a solution in phosphate buffered saline containing 250 mM imidizole and 50% glycerol.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service