Skip to Content
Merck
All Photos(1)

Key Documents

B1186

Sigma-Aldrich

Binucleine 2

≥97% (HPLC)

Synonym(s):

N′-(1-(3-Chloro-4-fluorophenyl)-4-cyano-1H-pyrazol-5-yl)-N,N-dimethyliminoformamide

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
$97.30

$97.30


Estimated to ship on29 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
2 MG
$97.30

About This Item

Empirical Formula (Hill Notation):
C13H11N5ClF
CAS Number:
Molecular Weight:
291.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$97.30


Estimated to ship on29 May 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥97% (HPLC)

form

solid

solubility

DMSO: soluble >5 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

CN(C)\C=N\c1c(cnn1-c2ccc(F)c(Cl)c2)C#N

InChI

1S/C13H11ClFN5/c1-19(2)8-17-13-9(6-16)7-18-20(13)10-3-4-12(15)11(14)5-10/h3-5,7-8H,1-2H3/b17-8+

InChI key

MNCYLHJHZOUDLB-CAOOACKPSA-N

Biochem/physiol Actions

Binucleine 2 is a cytokinesis inhibitor.
Cytokinesis inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chantal Roubinet et al.
Current biology : CB, 31(18), 3973-3983 (2021-07-24)
Although nuclei are the defining features of eukaryotes, we still do not fully understand how the nuclear compartment is duplicated and partitioned during division. This is especially the case for organisms that do not completely disassemble their nuclear envelope upon

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service