Skip to Content
Merck
All Photos(1)

Key Documents

B104

Sigma-Aldrich

tert-Butyl bicyclo[2.2.2]phosphorothionate

solid

Synonym(s):

TBPS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15O3PS
CAS Number:
Molecular Weight:
222.24
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

white to pink

solubility

DMSO: soluble
H2O: slightly soluble
ethanol: soluble

storage temp.

2-8°C

SMILES string

CC(C)(C)[C@@]12COP(=S)(OC1)OC2

InChI

1S/C8H15O3PS/c1-7(2,3)8-4-9-12(13,10-5-8)11-6-8/h4-6H2,1-3H3/t8-,12?

InChI key

VTBHBNXGFPTBJL-KRDXSZEZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aqueous solubility may be enhanced by the use of non-ionic detergents (Brij, TWEEN) or 2-hydroxypropyl-β-cyclodextrin.

Biochem/physiol Actions

GABAA receptor antagonist; chloride channel blocker; extremely potent convulsant.

Features and Benefits

This compound is featured on the GABAA Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

Solutions may be stored for 1-2 days at 4 °C.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alex S Evers et al.
The Journal of pharmacology and experimental therapeutics, 333(2), 404-413 (2010-02-04)
In the absence of GABA, neuroactive steroids that enhance GABA-mediated currents modulate binding of [35S]t-butylbicyclophosphorothionate in a biphasic manner, with enhancement of binding at low concentrations (site NS1) and inhibition at higher concentrations (site NS2). In the current study, compound
Daniel A García et al.
European journal of pharmacology, 600(1-3), 26-31 (2008-10-22)
Thymol is a monoterpene that specifically interacts with synaptic neural functions in neuronal GABA-operated Cl(-) channels. Here we explore the effects of thymol, and propofol as positive control, on t-[(35)S]butylbicyclophosphorothionate ([(35)S]TBPS) binding in primary cultures of cortical neurons. The study
Lu-Tai Tien et al.
Neurochemical research, 32(11), 1891-1897 (2007-06-15)
Anatomical evidence indicates that gamma-aminobutyric acid (GABA)-ergic and opioidergic systems are closely linked and act on the same neurons. However, the regulatory mechanisms between GABAergic and opioidergic system have not been well characterized. In the present study, we investigated whether
Shirin Strohmeier et al.
Pathogens (Basel, Switzerland), 8(4) (2019-10-24)
H11 subtype influenza viruses were isolated from a wide range of bird species and one strain also was isolated from swine. In an effort to generate reagents for a chimeric H11/1 hemagglutinin-based universal influenza virus vaccine candidate, we produced 28
J C Behrends
British journal of pharmacology, 129(2), 402-408 (2000-02-29)
1. T-butyl-bicyclo-phosphorothionate (TBPS) is a prototypical representative of the cage-convulsants which act through a use-dependent block of the GABA(A)-receptor-ionophore complex. Using current recordings from cultured neurones of rat striatum the manner was investigated in which two antagonists, bicuculline and penicillin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service