Skip to Content
Merck
All Photos(1)

Documents

B0684

Sigma-Aldrich

Anti-Bad antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-BBC2, Anti-BCL2L8

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse, human

technique(s)

western blot: 1 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BAD(572)
mouse ... Bad(12015)

Related Categories

General description

Bad (Bcl-2-antagonist of cell death), initially identified by its interaction with Bcl-2 and Bcl-XL, is a distant Bcl-2 family member. Bad bears only the most universally conserved amino acids within BH1 and BH2 domains, and lacks the typical hydrophobic C-terminal signal anchor.

Immunogen

synthetic peptide corresponding to amino acids 1-21 of human Bad conjugated to KLH.

Application

Anti-Bad antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL

Biochem/physiol Actions

The protein encoded by this gene forms heterodimers with BCL-xL and BCL-2 and replaces Bax, which in turn inverts the death repressor activity of Bcl-xL and positively regulates cell apoptosis.Bad is phosphorylated on two serine residues embedded in canonical 14-3-3 binding sites in response to IL-3, a survival factor. Phosphorylated Bad does not bind Bcl-XL and is sequestered in the cytosol bound to 14-3-3, a specific phosphoserine-binding protein. The growth factors that promote cell survival activate the threonine kinase Akt, which phosphorylates Bad causing suppression of apoptosis.

Physical form

lyophilized from a 0.2 μm filtered solution of phosphate buffered saline (PBS) and 5% trehalose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lishuai Feng et al.
International journal of molecular medicine, 42(2), 1074-1085 (2018-05-12)
Mono‑unsaturated free fatty acids (FFAs) can serve as a predictive indicator of vascular restenosis following interventional therapy, particularly in individuals with high‑fat diet‑induced type 2 diabetes. However, the pathogenic mechanism remains to be fully elucidated. In the present study, the
E Yang et al.
Cell, 80(2), 285-291 (1995-01-27)
To extend the mammalian cell death pathway, we screened for further Bcl-2 interacting proteins. Both yeast two-hybrid screening and lambda expression cloning identified a novel interacting protein, Bad, whose homology to Bcl-2 is limited to the BH1 and BH2 domains.
K Wang et al.
Molecular and cellular biology, 18(10), 6083-6089 (1998-09-19)
The BCL-2 family of proteins is comprised of proapoptotic as well as antiapoptotic members (S. N. Farrow and R. Brown, Curr. Opin. Genet. Dev. 6:45-49, 1996). A prominent death agonist, BAX, forms homodimers and heterodimerizes with multiple antiapoptotic members. Death
Z N Oltvai et al.
Cell, 74(4), 609-619 (1993-08-27)
Bcl-2 protein is able to repress a number of apoptotic death programs. To investigate the mechanism of Bcl-2's effect, we examined whether Bcl-2 interacted with other proteins. We identified an associated 21 kd protein partner, Bax, that has extensive amino

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service