Skip to Content
Merck
All Photos(1)

Key Documents

AXYZT50R

Pipette tips for Zymark® automated liquid handling system

size 50 μL, pkg of 5x960tips/cs, clear, racked

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
size:
50 μL
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc ZT-50-R
Corning
Product AXYZT50R is not currently sold in your country. Contact Technical Service

manufacturer/tradename

Axygen Inc ZT-50-R
Corning

packaging

pkg of 5x960tips/cs

size

50 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Legal Information

Zymark is a registered trademark of Sotax Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service