Skip to Content
Merck
All Photos(1)

Key Documents

AXYTF420LRS

MAXYMum Recovery® pipette filter tips

for Eppendorf® pipettors, ultramicro, size 0.5-20 μL, pkg of 5x960tips/cs, sterile

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.02
volume:
0.5—20 μL
sterility:
sterile
feature:
DNA free
RNase free
barrier: no
endotoxin free
filter
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc TF-420-L-R-S
Corning
Product AXYTF420LRS is not currently sold in your country. Contact Technical Service

material

clear

sterility

sterile

feature

DNA free
RNase free
barrier: no
endotoxin free
filter

manufacturer/tradename

Axygen Inc TF-420-L-R-S
Corning

packaging

pkg of 5x960tips/cs

size

0.5-20 μL

volume

0.5—20 μL

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Maxymum Recovery is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service