AXYTF200RS
Pipette filter tips
universal fit, racked, size 200 μL, pkg of 5x960tips/cs, sterile
Synonym(s):
Pipette tips, pipette tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
clear
sterility
sterile
feature
DNA free
RNase free
barrier
endotoxin free
filter
manufacturer/tradename
Axygen Inc TF-200-R-S
Corning
packaging
pkg of 5x960tips/cs
size
200 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Axygen® Filter Barrier tips feature a filter that creates a physical barrier by blocking aerosols and effectively preventing cross-contamination between samples. Additionally, it contains robotically inserted polyethylene filters to prevent nuclease contamination and are packaged in sealed-bottom racks with lids to maintain sterility and prevent external contamination. They are a reliable choice in situations where even minimal contamination can hamper results.
Application
Axygen® Filter Barrier tips have been used in microbiological and DNA amplification processes.
Features and Benefits
- These filter tips come in the sterilized form and are stored in sealed-bottom racks with lids to avoid contamination
- The filter creates a barrier within the tip to hinder and block aerosols, preventing cross-contamination between samples
- To prevent nuclease contamination, each polyethylene filter is robotically put into the tip and is devoid of DNA inhibitors
- Nonpyrogenic
- RNase-/DNase-free
Other Notes
Browse all pipette tips
Legal Information
Axygen is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Pathogenicity Assay of Penicillium expansum on Apple Fruits
Bio-protocol (2017)
Formation of Minimised Hairpin Template-transcribing Dumbbell Vectors for Small RNA Expression
Bio-protocol (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service