Skip to Content
Merck
All Photos(1)

Key Documents

AXYTF20

Pipette filter tips

for P-20, size 20 μL, pkg of 10x1000tips/cs

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.02
volume:
20 μL
sterility:
non-sterile
feature:
DNA free
RNase free
barrier
endotoxin free
filter
packaging:
pkg of 10x1000tips/cs
manufacturer/tradename:
Axygen Inc TF-20
Corning
Product AXYTF20 is not currently sold in your country. Contact Technical Service

material

clear

sterility

non-sterile

feature

DNA free
RNase free
barrier
endotoxin free
filter

manufacturer/tradename

Axygen Inc TF-20
Corning

packaging

pkg of 10x1000tips/cs

size

20 μL

volume

20 μL

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Pipette filter tips have been used in microbiological and DNA amplification processes.

Features and Benefits

  • Comes in the sterilized form and is stored in sealed-bottom racks with lids to avoid contamination
  • To prevent nuclease contamination, each polyethylene filter is robotically put into the tip and is devoid of DNA inhibitors
  • Filter creates a barrier within the tip to hinder and block aerosols, preventing cross-contamination between samples
  • Nonpyrogenic
  • RNase-/DNase-free

Other Notes

Browse all pipette tips

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service