Skip to Content
Merck
All Photos(1)

Key Documents

AXYTF1000

Pipette filter tips

universal fit, size 1000 μL, pkg of 5x1000tips/cs

Synonym(s):

Pipette tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.02
volume:
1000 μL
sterility:
non-sterile
feature:
DNA free
RNase free
barrier
endotoxin free
filter
packaging:
pkg of 5x1000tips/cs
manufacturer/tradename:
Axygen Inc TF-1000
Corning
Product AXYTF1000 is not currently sold in your country. Contact Technical Service

material

clear

sterility

non-sterile

feature

DNA free
RNase free
barrier
endotoxin free
filter

manufacturer/tradename

Axygen Inc TF-1000
Corning

packaging

pkg of 5x1000tips/cs

size

1000 μL

volume

1000 μL

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Pipette filter tips are suitable for microbiological and DNA amplification processes.

Features and Benefits

  • Sterile tips are stored in sealed-bottom racks with lids
  • The filter forms a barrier within the tip to prevent and block aerosols, avoiding cross-contamination between samples
  • Each polyethylene filter lacking DNA inhibitors, is robotically put into the tip to prevent nuclease contamination
  • Nonpyrogenic

Other Notes

Browse all pipette tips

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service