AXYT400L
MAXYMum Recovery® ultra micro pipette tips
for Eppendorf®, size 0.5-10 μL, pkg of 20x1000tips/cs
Synonym(s):
pipet tips
About This Item
RNase free
barrier: no
endotoxin free
filter: no
Corning
Recommended Products
material
clear
sterility
non-sterile
feature
DNA free
RNase free
barrier: no
endotoxin free
filter: no
manufacturer/tradename
Axygen Inc T-400-L
Corning
packaging
pkg of 20x1000tips/cs
size
0.5-10 μL
Looking for similar products? Visit Product Comparison Guide
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service