AXYSCT200SSCS
Screw cap tube
self-standing, size 2.0 mL, clear, pkg of 8x500caps/cs, sterile
Synonym(s):
96 multiwell plates, 96 well micoplates, 96 well plates, Corning tubes, cap tubes, polypropylene tubes
About This Item
Recommended Products
material
conical bottom tube
polypropylene
screw top cap
sterility
sterile
manufacturer/tradename
Axygen Inc SCT-200-SS-C-S
Corning
packaging
pkg of 8x500caps/cs
parameter
20000 × g max. RCF
size
2.0 mL
color
clear
clear
fitting
cap (screw cap)
suitability
suitable for (centrifugation)
Looking for similar products? Visit Product Comparison Guide
General description
- Axygen® microcentrifuge tubes with screw caps ensure a perfectly secure seal for sample protection.
- Screw caps with O-Rings to prevent sample evaporation.
- Free of RNase-/DNase and Nonpyrogenic.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service