Skip to Content
Merck
All Photos(1)

Key Documents

AXYSCT200SSB

Screw cap tubes and caps with O-rings, self-standing

size 2.0 mL, blue, pkg of 8x500caps/cs

Synonym(s):

centrifuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
Product AXYSCT200SSB is not currently sold in your country. Contact Technical Service

manufacturer/tradename

Axygen Inc SCT-200-SS-B
Corning

packaging

pkg of 8x500caps/cs

size

2.0 mL

color

blue

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service