Key Documents
AXYSCT150GS
Screw cap tubes and caps with O-rings, conical bottom
capacity 1.5 mL, green, sterile
Synonym(s):
centrifuge tubes
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
Product AXYSCT150GS is not currently sold in your country. Contact Technical Service
material
green
screw top cap
sterility
sterile
feature
RCF 20,000 × g
packaging
pack of 500 tubes
case of 8 packs
manufacturer/tradename
Axygen Inc
Corning
capacity
1.5 mL
Choose from one of the most recent versions:
Certificates of Analysis (COA)