AXYSCT050SSGRS
Self standing screw cap tubes and attached caps with O-rings
capacity 0.5 mL, gray, sterile
Synonym(s):
centrifuge tubes
All Photos(1)
About This Item
UNSPSC Code:
41121800
material
gray
screw top cap
sterility
sterile
feature
RCF 20,000 × g
packaging
pack of 500 tubes
case of 8 packs
manufacturer/tradename
Axygen Inc
Corning
capacity
0.5 mL
Choose from one of the most recent versions:
Certificates of Analysis (COA)