AXYPDW20C
Deep well plate
size 96 wells, size 2.0 mL, pkg of 10x5plates/cs
Synonym(s):
Pipette tips for Velocity
About This Item
Recommended Products
material
clear bottom
round bottom wells
sterility
non-sterile
feature
lid: no
skirt
manufacturer/tradename
Axygen Inc P-DW-20-C
Corning
packaging
pkg of 10x5plates/cs
size
2.0 mL
96 wells
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Axygen® deep well and assay microplates are made of chemically resistant and temperature resistant high-quality polypropylene.
- Ideal for sample collection, invitro growth chamber, and long-term storage.
- The round-well design of the microplate ensures complete sample recovery.
- The plates can be sealed using a heat-sealing film for sample protection.
- Axygen® deep well assay plates are ideal for use in applications including sample collection, in vitro growth chambers and long-term storage.
- Chemically resistant and withstands temperatures down to -80°C.
- Ultra-low profile design for reduced space requirements.
- Flat surface for efficient sealing using heat sealing film.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service