Skip to Content
Merck
All Photos(1)

Key Documents

AXYPCR384LC480WNF

PCR microplate for Roche® 480 LightCycler®

no film, size 384 wells, white, pkg of 5x10plates/cs

Synonym(s):

PCR microplate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
conical bottom white polypropylene wells
size:
384 wells
Product AXYPCR384LC480WNF is not currently sold in your country. Contact Technical Service

material

conical bottom white polypropylene wells

sterility

non-sterile

feature

lid: no
skirt Full skirt

manufacturer/tradename

Axygen Inc PCR-384-LC480-W-NF
Corning

packaging

pkg of 5x10plates/cs

maximum volume

30 μL

size

384 wells

working volume

30 μL

color

white

suitability

suitable for PCR

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.
LightCycler is a registered trademark of Roche
Roche is a registered trademark of Roche Molecular Systems, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service