Skip to Content
Merck
All Photos(1)

Key Documents

AXYPCR02DLC

MAXYMum Recovery® PCR tubes

with domed caps, thin wall, size 0.2 mL, pkg of 10x1000tubes/cs

Synonym(s):

centrifuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
Product AXYPCR02DLC is not currently sold in your country. Contact Technical Service

material

cap (snap cap)
conical bottom tube
polypropylene
polypropylene cap

sterility

non-sterile

manufacturer/tradename

Axygen Inc PCR-02D-L-C
Corning

packaging

pkg of 10x1000tubes/cs

size

0.2 mL

color

clear

suitability

suitable for PCR

binding type

(ultra-low retention)

Looking for similar products? Visit Product Comparison Guide

Legal Information

Maxymum Recovery is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service