Skip to Content
Merck
All Photos(1)

Key Documents

AXYMTXF1250CRS

Matrix pipette filter tips, 1250 μL, racked, sterile

pkg of 3x1152tips/cs

Synonym(s):

1250 μL Filter Tips, Filter Tips for Pipetting, Matrix Pipette Tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.02
volume:
1250 μL
sterility:
sterile
packaging:
pkg of 3x1152tips/cs
manufacturer/tradename:
Axygen Inc MTXF-1250-C-R-S
Corning
Product AXYMTXF1250CRS is not currently sold in your country. Contact Technical Service

sterility

sterile

manufacturer/tradename

Axygen Inc MTXF-1250-C-R-S
Corning

packaging

pkg of 3x1152tips/cs

capacity

1250 μL

volume

1250 μL

Other Notes

Browse all pipette tips

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service