Skip to Content
Merck
All Photos(1)

Key Documents

AXYMCT150X

Boil proof microtubes

size 1.5 mL, amber, pkg of 10x500tubes/cs

Synonym(s):

centrifuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121702
NACRES:
NB.22
Product AXYMCT150X is not currently sold in your country. Contact Technical Service

material

cap (snap cap)
conical bottom tube
polypropylene
polypropylene cap

sterility

non-sterile

feature

RNase and DNase free
endotoxin free
graduations

manufacturer/tradename

Axygen Inc MCT-150-X
Corning

packaging

pkg of 10x500tubes/cs

size

1.5 mL

color

amber

fitting

cap (push fit)

suitability

suitable for (centrifugation)

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service