Key Documents
AXYAM750ULRD
Silicone sealing mat for minitubes, pierceable, 96 Elisa and storage plates, and 750 μL 96 well plates
pkg of 5x10mats/cs
About This Item
UNSPSC Code:
41122410
NACRES:
NB.22
Product AXYAM750ULRD is not currently sold in your country. Contact Technical Service
material
silicone (research-grade)
sterility
non-sterile
manufacturer/tradename
Axygen Inc AM-750UL-RD
Corning
packaging
pkg of 5x10mats/cs
General description
  • Axygen® silicone sealing mat is ideal for use with 96 round well deep-well plates and 96-well ELISA plates.
  • Sealing mats reduce evaporation in PCR and storage applications
  • Made of autoclavable research-grade silicone
  • Reusable by following a bleach wash and ethanol rinse protocol.
  • Optimized to fit Axygen® MTS system, 1.0 mL Greiner and Corning deep well plates.
Legal Information
Axygen is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)