Key Documents
AXYAM384PCRRD
Sealing mat
for PCR microplates, size 384 wells, pkg of 5x10mats/cs
About This Item
UNSPSC Code:
41122410
NACRES:
NB.22
Product AXYAM384PCRRD is not currently sold in your country. Contact Technical Service
material
polypropylene
sterility
non-sterile
manufacturer/tradename
Axygen Inc AM-384-PCR-RD
Corning
packaging
pkg of 5x10mats/cs
size
384 wells
General description
  • Axygen® sealing films are used in PCR storage applications.
  • Sealing films offer high-temperature resistance.
  • Made from autoclavable research-grade silicone
  • Reusable - following a bleach wash and ethanol rinse protocol.
Legal Information
Axygen is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)