Skip to Content
Merck
All Photos(1)

Documents

AV54274

Sigma-Aldrich

Anti-ATG10 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-APG10, Anti-APG10L, Anti-ATG10 autophagy related 10 homolog (S. cerevisiae), Anti-DKFZP586I0418, Anti-FLJ13954, Anti-Pp12616

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

25 kDa

species reactivity

mouse, rat, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATG10(83734)

Related Categories

General description

ATG10 [autophagy related 10 homolog (S. cerevisiae)] or APG10 gene encodes a protein necessary for autophagy in yeast. Autophagy is a catabolic cellular event that includes cell death of superfluous or dysfunctional cellular components, cell differentiation and aging. It is also crucial for the maintenance of amino acid levels and protein synthesis under nitrogen starvation and is enhanced under certain conditions such as hormonal stimulation and drug treatments.

Immunogen

Synthetic peptide directed towards the C terminal region of human ATG10

Application

Anti-ATG10 antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL.

Biochem/physiol Actions

Atg10 is an E2-like enzyme that facilitates the addition of ubiquitination modifications essential for autophagosome formation. It catalyzes the conjugation of ATG12 to ATG5, which is essential for proper localization of ATG8 to the preautophagosomal structure (PAS). Further, Atg10 also plays a role in modifying the soluble form of LC3 to the membrane-bound form during Atg12 conjugation.

Sequence

Synthetic peptide located within the following region: TPVLKNSQKINKNVNYITSWLSIVGPVVGLNLPLSYAKATSQDERNVP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[The nurse; cancerology and radiotherapy].
C Chenal et al.
Revue de l'infirmiere, 26(8), 686-695 (1976-10-01)
Miao-Qing Zhang et al.
Frontiers in immunology, 9, 2176-2176 (2018-10-16)
Autophagy-related 10 (ATG10) is essential for autophagy since it promotes ATG5-ATG12 complex formation. Our previous study found that there are two isoforms of the ATG10 protein, ATG10 (a longer one) and ATG10S, which have identical sequences except an absence of
N Mizushima et al.
Nature, 395(6700), 395-398 (1998-10-06)
Autophagy is a process for the bulk degradation of proteins, in which cytoplasmic components of the cell are enclosed by double-membrane structures known as autophagosomes for delivery to lysosomes or vacuoles for degradation. This process is crucial for survival during
Takahiro Nemoto et al.
The Journal of biological chemistry, 278(41), 39517-39526 (2003-08-02)
Autophagy is a process for the bulk degradation of cytosolic compartments by lysosomes/vacuoles. The formation of autophagosomes involves a dynamic rearrangement of the membrane for which two ubiquitin-like modifications (the conjugation of Apg12p and the modification of a soluble form

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service