Skip to Content
Merck
All Photos(2)

Key Documents

AV50850

Sigma-Aldrich

Anti-WDFY3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ALFY, Anti-KIAA0993, Anti-MGC16461, Anti-WD repeat and FYVE domain containing 3, Anti-ZFYVE25

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$716.00

$716.00


Estimated to ship on21 May 2025



Select a Size

Change View
100 μL
$716.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$716.00


Estimated to ship on21 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

90 kDa

species reactivity

bovine, human, pig, rabbit, dog, mouse, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WDFY3(23001)

Immunogen

Synthetic peptide directed towards the C terminal region of human WDFY3

Application

Anti-WDFY3 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

WD repeat and FYVE domain containing 3 (WDFY3) is a phosphatidylinositol 3-phosphate-binding protein that shuttles from nuclear membrane and forms complexes with aggregated proteins. It is important for clearance of aggregate proteins and induction of autophagy.

Sequence

Synthetic peptide located within the following region: EKLADAVRFLGCFSDLRKISAMNVFPSNTQPFQRLLEEDVISIESVSPTL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Isakson et al.
Cell death and differentiation, 20(1), 12-20 (2012-06-02)
Autophagy, a highly conserved lysosomal degradation pathway, was initially characterized as a bulk degradation system induced in response to starvation. In recent years, autophagy has emerged also as a highly selective pathway, targeting various cargoes such as aggregated proteins and
Terje Høyvarde Clausen et al.
Autophagy, 6(3), 330-344 (2010-02-20)
Accumulation of ubiquitinated proteins in cytoplasmic and/or nuclear inclusions is a hallmark of several diseases associated with premature cell death. SQSTM1/p62 is known to bind ubiquitinated substrates and aid their aggregation and degradation by macroautophagy. We show here that p62
Tsung Lin Tsai et al.
Oncotarget, 8(53), 91209-91222 (2017-12-07)
Wheat germ agglutinin (WGA) is a lectin that specifically binds cell surface glycoproteins and disrupts nuclear pore complex function through its interaction with POM121. Our data indicate WGA induces paraptosis-like cell death without caspase activation. We observed the main features

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service