Skip to Content
Merck
All Photos(1)

Key Documents

AV50555

Sigma-Aldrich

Anti-CASP8AP2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CASP8 associated protein 2, Anti-CED-4, Anti-FLASH, Anti-FLJ11208, Anti-KIAA1315, Anti-RIP25

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$537.00

$537.00


Check Cart for Availability


Select a Size

Change View
100 μL
$537.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$537.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

223 kDa

species reactivity

human, rat, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CASP8AP2(9994)

General description

CASP8AP2 codes for caspase 8 associated protein 2 and it may be implicated in apoptosis. This gene has been studied as a prognostic biomarker in pediatric acute lymphoblastic leukemia.
Rabbit Anti-CASP8AP2 antibody recognizes canine, bovine, human, mouse, and rat CASSP8AP2.

Immunogen

Synthetic peptide directed towards the N terminal region of human CASP8AP2

Application

Rabbit Anti-CASP8AP2 antibody is suitable for western blot applications at a concentration of 1μg/ml.

Biochem/physiol Actions

CASP8AP2 is highly similar to FLASH, a mouse apoptotic protein identified by its interaction with the death-effector domain (DED) of caspase 8. Studies of FLASH protein suggested that this protein may be a component of the death-inducing signaling complex that includes Fas receptor, Fas-binding adapter FADD, and caspase 8, and plays a regulatory role in Fas-mediated apoptosis.This protein is highly similar to FLASH, a mouse apoptotic protein identified by its interaction with the death-effector domain (DED) of caspase 8. Studies of FLASH protein suggested that this protein may be a component of the death-inducing signaling complex that includes Fas receptor, Fas-binding adapter FADD, and caspase 8, and plays a regulatory role in Fas-mediated apoptosis. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: SLKKNISALIKTARVEINRKDEEISNLHQRLSEFPHFRNNHKTARTFDTV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Jiao et al.
Leukemia research, 36(1), 67-71 (2011-06-24)
The prognostic significance of caspase 8 associated protein 2 (CASP8AP2) in pediatric ALL is controversial. We determined a cut-off of CASP8AP2 expression in bone marrow samples of 39 newly diagnosed patients, and found a significantly poor bone marrow relapse-free survival

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service