Skip to Content
Merck
All Photos(2)

Key Documents

AV49500

Sigma-Aldrich

Anti-PLXDC1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp686F0937, Anti-FLJ36270, Anti-FLJ45632, Anti-Plexin domain containing 1, Anti-TEM3, Anti-TEM7

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$716.00

$716.00


Estimated to ship on29 May 2025



Select a Size

Change View
100 μL
$716.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$716.00


Estimated to ship on29 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

54 kDa

species reactivity

dog, human, rat, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLXDC1(57125)

Immunogen

Synthetic peptide directed towards the N terminal region of human PLXDC1

Application

Anti-PLXDC1 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

Plexin domain containing 1 (PLXDC1; tumor endothelial marker 7, TEM7) is a transmembrane and secreted protein containing plexin-like and weak nidogen-like domains. It mediates protein-protein interactions and is associated with angiogenic state of these cells. It is highly expressed in endothelium of tumor cells in a variety of cancer types.

Sequence

Synthetic peptide located within the following region: MDTLPDNRTRVVEDNHSYYVSRLYGPSEPHSRELWVDVAEANRSQVKIHT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Judy R van Beijnum et al.
Frontiers in bioscience (Scholar edition), 1, 216-225 (2009-06-02)
Endothelial cells (EC) are attractive targets for therapeutic interference in diseases that are dependent on the formation of novel blood vessels, such as cancer. EC are readily accessible via the blood stream and are considered to be genetically stable, thus
Rebecca G Bagley et al.
Microvascular research, 82(3), 253-262 (2011-10-01)
Antiangiogenesis has been validated as a therapeutic strategy to treat cancer, however, a need remains to identify new targets and therapies for specific diseases and to improve clinical benefit from antiangiogenic agents. Tumor endothelial marker 7 (TEM-7) was investigated as
Akash Nanda et al.
Cancer research, 64(23), 8507-8511 (2004-12-03)
Tumor endothelial marker 7 (TEM7) was recently identified as an mRNA transcript overexpressed in the blood vessels of human solid tumors. Here, we identify several new variants of TEM7, derived by alternative splicing, that are predicted to be intracellular (TEM7-I)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service