Skip to Content
Merck
All Photos(2)

Key Documents

AV48481

Sigma-Aldrich

Anti-OXCT1 (AB2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-3-Oxoacid CoA transferase 1, Anti-OXCT, Anti-SCOT

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$537.00

$537.00


Estimated to ship on21 April 2025



Select a Size

Change View
100 μL
$537.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$537.00


Estimated to ship on21 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

52 kDa

species reactivity

yeast, guinea pig, rat, bovine, human, mouse, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OXCT1(5019)

General description

3-Oxoacid CoA transferase 1 (OCT1) is a mitochondrial enzyme that catalyzes the transfer of coenzyme A from succinyl-CoA to acetoacetate during ketone body catabolism. Mutations in OXCT1 have been linked to Succinyl-CoA:3-ketoacid CoA transferase (SCOT) deficiency.
Rabbit Anti-OXCT1 antibody recognizes pig, bovine, chicken, human, mouse, and rat, OXCT1.

Immunogen

Synthetic peptide directed towards the middle region of human OXCT1

Application

Rabbit Anti-OXCT1 (AB2) antibody is suitable for western blot applications at a concentration of 1 μg/ml.

Biochem/physiol Actions

CT1 is a member of the 3-oxoacid CoA-transferase gene family. It is a homodimeric mitochondrial matrix enzyme that plays a central role in extrahepatic ketone body catabolism by catalyzing the reversible transfer of coenzyme A from succinyl-CoA to acetoacetate.This gene encodes a member of the 3-oxoacid CoA-transferase gene family. The encoded protein is a homodimeric mitochondrial matrix enzyme that plays a central role in extrahepatic ketone body catabolism by catalyzing the reversible transfer of coenzyme A from succinyl-CoA to acetoacetate. Mutations in this gene are associated with succinyl CoA:3-oxoacid CoA transferase deficiency.This gene encodes a member of the 3-oxoacid CoA-transferase gene family. The encoded protein is a homodimeric mitochondrial matrix enzyme that plays a central role in extrahepatic ketone body catabolism by catalyzing the reversible transfer of coenzyme A from succinyl-CoA to acetoacetate. Mutations in this gene are associated with succinyl CoA:3-oxoacid CoA transferase deficiency.

Sequence

Synthetic peptide located within the following region: GMYANLGIGIPLLASNFISPNITVHLQSENGVLGLGPYPRQHEADADLIN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshiyuki Fukao et al.
Journal of inherited metabolic disease, 33 Suppl 3, S307-S313 (2010-07-24)
Succinyl-CoA:3-ketoacid CoA transferase (SCOT) deficiency causes episodic ketoacidotic crises and no apparent symptoms between them. Here, we report a Japanese case of neonatal-onset SCOT deficiency. The male patient presented a severe ketoacidotic crisis, with blood pH of 7.072 and bicarbonate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service