Skip to Content
Merck
All Photos(1)

Key Documents

AV48273

Sigma-Aldrich

Anti-ALDOC antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ALDC, Anti-Aldolase C, fructose-bisphosphate

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$609.00

$609.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1354


Select a Size

Change View
100 μL
$609.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$609.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1354

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39 kDa

species reactivity

dog, rat, human, bovine, horse, guinea pig, rabbit, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALDOC(230)

General description

ALDOC codes for a class I fructose-biphosphate aldolase that catalyzes the breakdown of fructose-1,6-biphosphate to dihydroxyacetone phosphate (DHAP) and glyceraldehyde-3-phosphate during glycolysis. It also catalyzes the aldol cleavage of fructose-1 phosphate to DHAP and glyceraldehyde. ALDOC is expressed in the cerebellum, retina and other parts of the CNS. It is known to be upregulated in the brain and skeletal muscles cells of chickens during hypoxia.
Rabbit Anti-ALDOC antibody recognizes bovine, human, mouse, rat, zebrafish, and rabbit ALDOC.

Immunogen

Synthetic peptide directed towards the N terminal region of human ALDOC

Application

Rabbit Anti-ALDOC antibody is suitable for western blot applications at a concentration of 0.5 μg/ml.

Biochem/physiol Actions

ALDOC gene is a member of the class I fructose-biphosphate aldolase gene family. ALDOC is a glycolytic enzyme that catalyzes the reversible aldol cleavage of fructose-1,6-biphosphate and fructose 1-phosphate to dihydroxyacetone phosphate and either glyceraldehyde-3-phosphate or glyceraldehyde, respectively.This gene encodes a member of the class I fructose-biphosphate aldolase gene family. Expressed specifically in the hippocampus and Purkinje cells of the brain, the encoded protein is a glycolytic enzyme that catalyzes the reversible aldol cleavage of fructose-1,6-biphosphate and fructose 1-phosphate to dihydroxyacetone phosphate and either glyceraldehyde-3-phosphate or glyceraldehyde, respectively.

Sequence

Synthetic peptide located within the following region: MPHSYPALSAEQKKELSDIALRIVAPGKGILAADESVGSMAKRLSQIGVE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C F Wang et al.
Animal genetics, 38(3), 203-210 (2007-06-02)
Two sequence variants of the aldolase C (ALDOC) gene were discovered based on comparison of the sequences from an altiplano chicken breed (Tibetan chicken) and two lowland breeds (White Leghorn and ShouGuang). Gel-shift results indicated that one of these variants
Ryota Maruyama et al.
Pathology international, 72(3), 176-186 (2022-02-12)
Colorectal cancer (CRC) is a leading cause of cancer-related death worldwide. The spheroid colony formation assay is a useful method to identify cancer stem cells (CSCs). Using the DLD-1 and WiDr CRC cell lines, we performed microarray analyses of spheroid
Hirofumi Fujita et al.
PloS one, 9(1), e86679-e86679 (2014-01-30)
Aldolase C (Aldoc, also known as "zebrin II"), a brain type isozyme of a glycolysis enzyme, is expressed heterogeneously in subpopulations of cerebellar Purkinje cells (PCs) that are arranged longitudinally in a complex striped pattern in the cerebellar cortex, a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service